MilliporeSigma
Analysis of Tim-3 as a therapeutic target in prostate cancer.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-08-15)
Yong-Rui Piao, Zhe-Hu Jin, Kui-Chang Yuan, Xuan-Shun Jin
ABSTRACT
T cell immunoglobulin domain and mucin domain-containing molecule 3 (Tim-3) is a newly discovered immunomodulatory, which plays an important role in immunity regulation. Recent evidence suggests that Tim-3 is differentially regulated in a variety of tumors and has a potential as a therapeutic target. The aim of this study was to investigate the effect of Tim-3 on the development of prostate cancer (PCa). Tim-3 expressing on peripheral CD4+ T and CD8+ T cells was analyzed by flow cytometry. The relationships between Tim-3 expression and clinicopathological features were analyzed. Immunohistochemical expression of Tim-3 was examined in our large numbers of paraffin-fixed prostate tissues. Flow cytometry revealed that expression of Tim-3 was significantly increased on both CD4+ and CD8+ T cells in PCa patients than that in benign prostate hyperplasia (BPH) patients. Also, the level of Tim-3 on CD4+ T cells was positively correlated with CD8+ T cells in patients. Further analyses revealed that the levels of Tim-3 on CD4+ T cells and CD8+ T cells exhibited different expression patterns in terms of localization depending on pathological category of PCa and metastasis. Immunohistochemical analysis revealed that positive staining of Tim-3 in PCa but little or no staining of Tim-3 was observed in BPH epithelium. Tim-3 may affect the development and progression of PCa, which may provide knowledge for using Tim-3 as a novel therapy for effective PCa management.
Sigma-Aldrich
MISSION® esiRNA, targeting human HAVCR2
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC