MilliporeSigma
  • Sperm-associated antigen 16 is a novel target of the humoral autoimmune response in multiple sclerosis.
Sperm-associated antigen 16 is a novel target of the humoral autoimmune response in multiple sclerosis.
Journal of immunology (Baltimore, Md. : 1950) (2014-08-03)
Laura de Bock, Klaartje Somers, Judith Fraussen, Jerome J A Hendriks, Jack van Horssen, Myrthe Rouwette, Niels Hellings, Luisa M Villar, José C Alvarez-Cermeño, Mercedes Espiño, Raymond Hupperts, Peter Jongen, Jan Damoiseaux, Marcel M Verbeek, Peter P De Deyn, Marie D'hooghe, Bart Van Wijmeersch, Piet Stinissen, Veerle Somers
ABSTRACT
We have previously identified eight novel autoantibody targets in the cerebrospinal fluid of multiple sclerosis (MS) patients, including sperm-associated Ag 16 (SPAG16). In the current study, we further investigated the autoantibody response against SPAG16-a protein with unknown function in the CNS-and its expression in MS pathology. Using isoelectric focusing, we detected SPAG16-specific oligoclonal bands in the cerebrospinal fluid of 5 of 23 MS patients (22%). Analysis of the anti-SPAG16 Ab reactivity in the plasma of a total of 531 donors using ELISA demonstrated significantly elevated anti-SPAG16 Ab levels (p = 0.002) in 32 of 153 MS patients (21%) compared with all other control groups with 95% specificity for the disease. To investigate the pathologic relevance of anti-SPAG16 Abs in vivo, anti-SPAG16 Abs were injected in mice with experimental autoimmune encephalomyelitis, resulting in a significant disease exacerbation. Finally, we demonstrated a consistent upregulation of SPAG16 in MS brain and experimental autoimmune encephalomyelitis spinal cord lesions, more specifically in reactive astrocytes. We conclude that SPAG16 is a novel autoantibody target in a subgroup of MS patients and in combination with other diagnostic criteria, elevated levels of anti-SPAG16 Abs could be used as a biomarker for diagnosis. Furthermore, the pathologic relevance of anti-SPAG16 Abs was shown in vivo.
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®
Sigma-Aldrich
Hematoxylin, certified by the BSC