MilliporeSigma
  • Understanding and managing the impact of HPMC variability on drug release from controlled release formulations.
Understanding and managing the impact of HPMC variability on drug release from controlled release formulations.
Journal of pharmaceutical sciences (2014-03-22)
Deliang Zhou, Devalina Law, Judie Reynolds, Lynn Davis, Clifford Smith, Jose L Torres, Viraj Dave, Nishanth Gopinathan, Daniel T Hernandez, Mary Kay Springman, Casey Chun Zhou
ABSTRACT
The purpose of this study is to identify critical physicochemical properties of hydroxypxropyl methylcellulose (HPMC) that impact the dissolution of a controlled release tablet and develop a strategy to mitigate the HPMC lot-to-lot and vendor-to-vendor variability. A screening experiment was performed to evaluate the impacts of methoxy/hydroxypropyl substitutions, and viscosity on drug release. The chemical diversity of HPMC was explored by nuclear magnetic resonance (NMR), and the erosion rate of HPMC was investigated using various dissolution apparatuses. Statistical evaluation suggested that the hydroxypropyl content was the primary factor impacting the drug release. However, the statistical model prediction was not robust. NMR experiments suggested the existence of structural diversity of HPMC between lots and more significantly between vendors. Review of drug release from hydrophilic matrices indicated that erosion is a key aspect for both poorly soluble and soluble drugs. An erosion rate method was then developed, which enabled the establishment of a robust model and a meaningful HPMC specification. The study revealed that the overall substitution level is not the unique parameter that dictates its release-controlling properties. Fundamental principles of polymer chemistry and dissolution mechanisms are important in the development and manufacturing of hydrophilic matrices with consistent dissolution performance.
Product Description
Stearic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
USP
Stearic acid, United States Pharmacopeia (USP) Reference Standard