MilliporeSigma
  • Soluble toll-like receptor 2 is significantly elevated in HIV-1 infected breast milk and inhibits HIV-1 induced cellular activation, inflammation and infection.
Soluble toll-like receptor 2 is significantly elevated in HIV-1 infected breast milk and inhibits HIV-1 induced cellular activation, inflammation and infection.
AIDS (London, England) (2014-09-30)
Bethany M Henrick, Xiao-Dan Yao, Anna G Drannik, Alash'le Abimiku, Kenneth L Rosenthal
ABSTRACT
We previously demonstrated that immunodepletion of soluble Toll-like receptor 2 (sTLR2) from human breast milk significantly increased HIV infection in vitro. The aims of this study were to characterize sTLR2 levels in breast milk from HIV-infected and uninfected women, and identify a mechanism by which sTLR2 inhibits HIV-induced cellular activation and infection. Blinded studies of breast milk from HIV-infected and uninfected Nigerian and Canadian women were evaluated for levels of sTLR2, proinflammatory cytokines and viral antigenemia. In-vitro experiments were conducted using cell lines to assess sTLR2 function in innate responses and effect on HIV infection. Breast milk from HIV-infected women showed significantly higher levels of sTLR2 than uninfected breast milk. Further, sTLR2 levels correlated with HIV-1 p24 and interleukin (IL)-15, thus suggesting a local innate compensatory response in the HIV-infected breast. Given the significantly higher levels of sTLR2 in breast milk from HIV-infected women, we next demonstrated that mammary epithelial cells and macrophages, which are prevalent in milk, produced significantly increased levels of sTLR2 following exposure to HIV-1 proteins p17, p24 and gp41 or the TLR2 ligand, Pam3CSK4. Our results also demonstrated that sTLR2 physically interacts with p17, p24 and gp41 and inhibits HIV-induced nuclear factor kappa-light-chain-enhancer of activated B cells activation, and inflammation. Importantly, binding of sTLR2 to HIV-1 proteins inhibited a TLR2-dependent increase in chemokine receptor 5 expression, thus resulting in significantly reduced HIV-1 infection. These results indicate novel mechanisms by which sTLR2 plays a critical role in inhibiting mother-to-child HIV transmission.
Product Description
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard