MilliporeSigma
  • Regulation of diacylglycerol acyltransferase 2 protein stability by gp78-associated endoplasmic-reticulum-associated degradation.
Regulation of diacylglycerol acyltransferase 2 protein stability by gp78-associated endoplasmic-reticulum-associated degradation.
The FEBS journal (2014-05-14)
Kwangman Choi, Hyeongki Kim, Hyunju Kang, So-Young Lee, Sang Jun Lee, Sung Hoon Back, Seo Hyun Lee, M Sun Kim, Jeong Eun Lee, Ju Young Park, Jiye Kim, Sunhong Kim, Jae-Hyung Song, Yura Choi, Suui Lee, Hyun-Jun Lee, Jong Heon Kim, Sungchan Cho
ABSTRACT
Triacylglycerol (TG) is the major form of stored energy in eukaryotic organisms and is synthesized by diacylglycerol acyltransferase (DGAT) in the endoplasmic reticulum (ER). DGAT2, one of the two DGAT enzymes, is barely detectable in cells, even though its mRNA transcripts are maintained at considerable levels. However, little is known about how DGAT2 expression is altered by protein stability. DGAT2 was highly unstable in cells and was rapidly degraded by proteasomes in an ubiquitin-dependent manner. Deletion mutation analysis identified transmembrane domain 1 (TMD1) as a protein degradation signal. TMD1 is also important for ER localization of DGAT2. Moreover, DGAT2 interacted with p97/VCP, a crucial component of the ER-associated degradation (ERAD) pathway, and polyubiquitinated DGAT2 accumulated following treatment with an ERAD inhibitor. Furthermore, gp78, an E3 ligase involved in ERAD, regulates the degradation of DGAT2 through direct interactions and ubiquitination. Consequently, the stabilization of DGAT2 increased the number of lipid droplets in hepatic cells. Therefore, DGAT2 is regulated by gp78-associated ERAD at the post-translational level.
Sigma-Aldrich
Calpeptin, ≥98% (HPLC)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium chloride, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Ammonium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Supelco
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)