MilliporeSigma
  • Human CD4+ CD39+ regulatory T cells produce adenosine upon co-expression of surface CD73 or contact with CD73+ exosomes or CD73+ cells.
Human CD4+ CD39+ regulatory T cells produce adenosine upon co-expression of surface CD73 or contact with CD73+ exosomes or CD73+ cells.
Clinical and experimental immunology (2014-04-23)
P J Schuler, Z Saze, C-S Hong, L Muller, D G Gillespie, D Cheng, M Harasymczuk, M Mandapathil, S Lang, E K Jackson, T L Whiteside
ABSTRACT
While murine CD4(+) CD39(+) regulatory T cells (T(reg)) co-express CD73 and hydrolyze exogenous (e) adenosine triphosphate (ATP) to immunosuppressive adenosine (ADO), surface co-expression of CD73 on human circulating CD4(+) CD39(+) T(reg) is rare. Therefore, the ability of human T(reg) to produce and utilize ADO for suppression remains unclear. Using mass spectrometry, we measured nucleoside production by subsets of human CD4(+) CD39(+) and CD4(+) CD39(-)CD73(+) T cells or CD19(+) B cells isolated from blood of 30 volunteers and 14 cancer patients. CD39 and CD73 expression was evaluated by flow cytometry, Western blots, confocal microscopy or reverse transcription-polymerase chain reaction (RT-PCR). Circulating CD4(+) CD39(+) T(reg) which hydrolyzed eATP to 5'-AMP contained few intracytoplasmic granules and had low CD73 mRNA levels. Only ∼1% of these T(reg) were CD39(+) CD73(+) . In contrast, CD4(+) CD39(neg) CD73(+) T cells contained numerous CD73(+) granules in the cytoplasm and strongly expressed surface CD73. In vitro-generated T(reg) (Tr1) and most B cells were CD39(+) CD73(+) . All these CD73(+) T cell subsets and B cells hydrolyzed 5'-AMP to ADO. Exosomes isolated from plasma of normal control (NC) or cancer patients carried enzymatically active CD39 and CD73(+) and, when supplied with eATP, hydrolyzed it to ADO. Only CD4(+) CD39(+) T(reg) co-incubated with CD4(+) CD73(+) T cells, B cells or CD39(+) CD73(+) exosomes produced ADO. Thus, contact with membrane-tethered CD73 was sufficient for ADO production by CD4(+) CD39(+) T(reg). In microenvironments containing CD4(+) CD73(+) T cells, B cells or CD39(+) CD73(+) exosomes, CD73 is readily available to CD4(+) CD39(+) CD73(neg) T(reg) for the production of immunosuppressive ADO.
Product Description
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
Trometamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Trizma® base, BioUltra, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline