MilliporeSigma
  • Pemetrexed induces S-phase arrest and apoptosis via a deregulated activation of Akt signaling pathway.
Pemetrexed induces S-phase arrest and apoptosis via a deregulated activation of Akt signaling pathway.
PloS one (2014-05-23)
Kun-Chieh Chen, Tsung-Ying Yang, Chun-Chi Wu, Chi-Chih Cheng, Shih-Lan Hsu, Hsiao-Wen Hung, Jian-Wei Chen, Gee-Chen Chang
ABSTRACT
Pemetrexed is approved for first-line and maintenance treatment of patients with advanced or metastatic non-small-cell lung cancer (NSCLC). The protein kinase Akt/protein kinase B is a well-known regulator of cell survival which is activated by pemetrexed, but its role in pemetrexed-mediated cell death and its molecular mechanisms are unclear. This study showed that stimulation with pemetrexed induced S-phase arrest and cell apoptosis and a parallel increase in sustained Akt phosphorylation and nuclear accumulation in the NSCLC A549 cell line. Inhibition of Akt expression by Akt specific siRNA blocked S-phase arrest and protected cells from apoptosis, indicating an unexpected proapoptotic role of Akt in the pemetrexed-mediated toxicity. Treatment of A549 cells with pharmacological inhibitors of phosphatidylinositol 3-kinase (PI3K), wortmannin and Ly294002, similarly inhibited pemetrexed-induced S-phase arrest and apoptosis and Akt phosphorylation, indicating that PI3K is an upstream mediator of Akt and is involved in pemetrexed-mediated cell death. Previously, we identified cyclin A-associated cyclin-dependent kinase 2 (Cdk2) as the principal kinase that was required for pemetrexed-induced S-phase arrest and apoptosis. The current study showed that inhibition of Akt function and expression by pharmacological inhibitors as well as Akt siRNA drastically inhibited cyclin A/Cdk2 activation. These pemetrexed-mediated biological and molecular events were also observed in a H1299 cell line. Overall, our results indicate that, in contrast to its normal prosurvival role, the activated Akt plays a proapoptotic role in pemetrexed-mediated S-phase arrest and cell death through a mechanism that involves Cdk2/cyclin A activation.
Product Description
L-Threonine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
SAFC
L-Threonine
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Akt1
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
MISSION® esiRNA, targeting human AKT1, RP11-982M15.2
Sigma-Aldrich
Propidium iodide solution