MilliporeSigma
  • Comparison of in vivo efficacy of different ocular lubricants in dry eye animal models.
Comparison of in vivo efficacy of different ocular lubricants in dry eye animal models.
Investigative ophthalmology & visual science (2014-05-02)
Xiaodong Zheng, Tomoko Goto, Yuichi Ohashi
ABSTRACT
To compare the efficacy of three types of ocular lubricants in protecting corneal epithelial cells in dry eye animal models. Ocular lubricants containing 0.1% or 0.3% sodium hyaluronate (SH), carboxymethylcellulose (CMC), or hydroxypropyl methylcellulose (HPMC) were tested. First, ocular lubricant containing 0.002% fluorescein was dropped onto the rabbit corneas. The fluorescein intensity as an index of retention was measured. Second, a rabbit dry eye model was made by holding the eye open with a speculum, and 50 μL of each ocular lubricant was dropped onto the cornea. After 3 hours, the corneas were stained with 1% methylene blue (MB), and the absorbance of MB was measured. Third, a rat dry eye model was treated with the ocular lubricants for 4 weeks, and the corneal fluorescein staining was scored. Eyes treated with physiological saline were used as controls. Finally, immunohistochemistry was used to analyze occludin, an epithelial barrier protein, in cultured human corneal epithelial cells pretreated with ocular lubricants and desiccated for 20 or 60 minutes. Our results showed that 0.3% SH had a significantly longer retention time than the other lubricants (all P < 0.01). The absorbance of MB was significantly lower in the 0.3% SH group. The corneas of rats exposed to 0.3% SH had significantly lower fluorescein staining scores. A significantly higher number of occludin-positive cells were found after exposure to 0.3% SH than other lubricants. Ocular lubricant containing 0.3% SH would be preferable to treat patients with dry eye syndrome.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Fluorescein, free acid
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Propidium iodide solution
Supelco
Urethane, analytical standard
Sigma-Aldrich
Urethane, ≥99%
Sigma-Aldrich
Urethane, ≥99.0% (GC)