MilliporeSigma
B-cell-intrinsic STAT6 signaling controls germinal center formation.
European journal of immunology (2014-04-30)
Adriana Turqueti-Neves, Manuel Otte, Olivia Prazeres da Costa, Uta E Höpken, Martin Lipp, Thorsten Buch, David Voehringer
ABSTRACT
Infection with helminths and exposure to antigens induce a strong type 2 immune response resulting in the secretion of the cytokines IL-4 and IL-13 by CD4(+) T cells and several innate cell types. IL-4 and IL-13 promote class switch recombination to IgG1 and IgE while their role for germinal center (GC) formation is poorly understood. We found a dramatic reduction in the numbers of GC B cells when investigating different type 2 immune responses in IL-4/IL-13-deficient mice. IL-4/IL-13 from T cells located outside B-cell follicles was sufficient for GC formation. We further revealed that IL-4/IL-13 acts directly on B cells for the formation of a robust GC response. The frequency of apoptotic GC B cells was not altered in the absence of IL-4/IL-13 and proliferation was even enhanced. However, deficiency of signal transducer and activator of transcription 6 signaling in B cells resulted in failure to downregulate the chemotactic receptor Gpr183 (Ebi2) and downregulation of this receptor has been shown to be essential for proper GC B-cell differentiation. Thus, T-cell-derived extrafollicular IL-4/IL-13 and signal transducer and activator of transcription 6-regulated genes in B cells play a critical role for orchestration of the GC response in type 2 immunity.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Brefeldin A, BioXtra, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Propidium iodide solution