MilliporeSigma
  • Increased rho-kinase activity in hypertensive patients with left ventricular hypertrophy.
Increased rho-kinase activity in hypertensive patients with left ventricular hypertrophy.
American journal of hypertension (2013-12-24)
Luigi Gabrielli, Jose L Winter, Ivan Godoy, Paul McNab, Ivonne Padilla, Samuel Cordova, Paola Rigotti, Ulises Novoa, Italo Mora, Lorena García, Maria P Ocaranza, Jorge E Jalil
ABSTRACT
There is experimental evidence on the role of Rho-kinase (ROCK) activation in cardiac hypertrophy but no information on its role in human hypertension and left ventricular hypertrophy (LVH). We hypothesized that ROCK activity is higher in hypertensive patients with LVH compared with hypertensive patients without LVH. We conducted a cross-sectional study comparing untreated hypertensive patients with (n = 41) and without LVH (n = 46) determined by echocardiography with a healthy normotensive control group (n = 51). Measurements included LV mass, dimensions, and function and ROCK activity determined in circulating leukocytes by measuring Western blot levels of phosphorylated to total myosin light chain phosphatase 1 (MYPT1-p/t). Compared with normotensive subjects, MYPT1-p/t was significantly increased by 4.5-fold in the hypertensive patients without LVH and by 9-fold in the hypertensive patients with LVH. Compared with the hypertension without LVH group, MYPT1-p/t was significantly increased by 2-fold in the hypertension with LVH gorup. In patients with eccentric LVH, the mean MYPT1-p/t ratio was significantly higher by 4-fold compared with hypertensive patients without eccentric LVH. Patients with an E/e' ratio ≥15 (n = 6) showed a higher MYPT1-p/t ratio (by 26%) compared with patients with a lower E/e' ratio (P ≤ 0.01). ROCK activity is higher in hypertensive patients with LVH compared with hypertensive patients without LVH, and it is further increased when eccentric LVH is present. Thus, in hypertension, ROCK activation is related to pathological cardiac remodeling and might have a role as an LVH marker.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)