MilliporeSigma
  • Comparison of proton channel, phagocyte oxidase, and respiratory burst levels between human eosinophil and neutrophil granulocytes.
Comparison of proton channel, phagocyte oxidase, and respiratory burst levels between human eosinophil and neutrophil granulocytes.
Free radical research (2014-07-06)
I Kovács, M Horváth, T Kovács, K Somogyi, L Tretter, M Geiszt, G L Petheő
ABSTRACT
Robust production of reactive oxygen species (ROS) by phagocyte NADPH oxidase (phox) during the respiratory burst (RB) is a characteristic feature of eosinophil and neutrophil granulocytes. In these cells the voltage-gated proton channel (Hv1) is now considered as an ancillary subunit of the phox needed for intense ROS production. Multiple sources reported that the expression of phox subunits and RB is more intensive in eosinophils than in neutrophils. In most of these studies the eosinophils were not isolated from healthy individuals, and a comparative analysis of Hv1 expression had never been carried out. We performed a systematic comparison of the levels of essential phox subunits, Hv1 expression and ROS producing capacity between eosinophils and neutrophils of healthy individuals. The expression of phox components was similar, whereas the amount of Hv1 was ∼ 10-fold greater in eosinophils. Furthermore, Hv1 expression correlated with Nox2 expression only in eosinophils. Additionally, in confocal microscopy experiments co-accumulation of Hv1 and Nox2 at the cell periphery was observed in resting eosinophils but not in neutrophils. While phorbol-12-myristate-13-acetate-induced peak extracellular ROS release was ∼ 1.7-fold greater in eosinophils, oxygen consumption studies indicated that the maximal intensity of the RB is only ∼ 1.4-fold greater in eosinophils. Our data reinforce that eosinophils, unlike neutrophils, generate ROS predominantly extracellularly. In contrast to previous works we have found that the two granulocyte types display very similar phox subunit expression and RB capacity. The large difference in Hv1 expression suggests that its support to intense ROS production is more important at the cell surface.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
SAFC
HEPES
SAFC
Sodium chloride solution, 5 M
SAFC
HEPES
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%