MilliporeSigma
  • Repair of the injured spinal cord by implantation of a synthetic degradable block copolymer in rat.
Repair of the injured spinal cord by implantation of a synthetic degradable block copolymer in rat.
Biomaterials (2014-05-13)
Vincent Pertici, Thomas Trimaille, Jérôme Laurin, Marie-Solenne Felix, Tanguy Marqueste, Brigitte Pettmann, Jean-Paul Chauvin, Didier Gigmes, Patrick Decherchi
ABSTRACT
The present study is designed to assess the properties of a new degradable PLA-b-PHEMA block copolymer hydrogel and its therapeutic effectiveness after implantation following a thoracic spinal cord hemisection on rats. Degradable characteristics and porous aspect of the scaffold are respectively analyzed by the evaluation of its mass loss and by electron microscopy. The biomaterial toxicity is measured through in vitro tests based on motoneuron survival and neurite growth on copolymer substrate. Functional measurements are assessed by the Basso, Beattie and Bresnahan (BBB) and the Dynamic Weight Bearing (DWB) tests during 8 weeks post-surgery. Histological analyses are achieved to evaluate the presence of blood vessels and axons, the density of the glial scar, the inflammatory reaction and the myelination at the lesion site and around it. The results indicate that the synthetic PLA-b-PHEMA block copolymer is a non-toxic and degradable biomaterial that provides support for regenerating axons and seems to limit scar tissue formation. Additionally, the implantation of the porous PLA-b-PHEMA scaffold enhances locomotor improvement. The observed functional recovery highlights the potential benefits of plain tissue engineering material, which can further be optimized by bioactive molecule functionalization or transplanted cell encapsulation.
Product Description
Supelco
Oxytetracycline, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Oxytetracycline, United States Pharmacopeia (USP) Reference Standard
Millipore
Oxytetra Selective Supplement, suitable for microbiology
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioUltra, ≥99.5% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sucrose, European Pharmacopoeia (EP) Reference Standard
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard