MilliporeSigma
  • Dual oxidase 2 in lung epithelia is essential for hyperoxia-induced acute lung injury in mice.
Dual oxidase 2 in lung epithelia is essential for hyperoxia-induced acute lung injury in mice.
Antioxidants & redox signaling (2014-04-29)
Min-Ji Kim, Jae-Chan Ryu, Younghee Kwon, Suhee Lee, Yun Soo Bae, Joo-Heon Yoon, Ji-Hwan Ryu
ABSTRACT
Acute lung injury (ALI) induced by excessive hyperoxia has been employed as a model of oxidative stress imitating acute respiratory distress syndrome. Under hyperoxic conditions, overloading quantities of reactive oxygen species (ROS) are generated in both lung epithelial and endothelial cells, leading to ALI. Some NADPH oxidase (NOX) family enzymes are responsible for hyperoxia-induced ROS generation in lung epithelial and endothelial cells. However, the molecular mechanisms of ROS production in type II alveolar epithelial cells (AECs) and ALI induced by hyperoxia are poorly understood. In this study, we show that dual oxidase 2 (DUOX2) is a key NOX enzyme that affects hyperoxia-induced ROS production, particularly in type II AECs, leading to lung injury. In DUOX2 mutant mice (DUOX2(thyd/thyd)) or mice in which DUOX2 expression is knocked down in the lungs, hyperoxia-induced ALI was significantly lower than in wild-type (WT) mice. DUOX2 was mainly expressed in type II AECs, but not endothelial cells, and hyperoxia-induced ROS production was markedly reduced in primary type II AECs isolated from DUOX2(thyd/thyd) mice. Furthermore, DUOX2-generated ROS are responsible for caspase-mediated cell death, inducing ERK and JNK phophorylation in type II AECs. To date, no role for DUOX2 has been defined in hyperoxia-mediated ALI despite it being a NOX homologue and major ROS source in lung epithelium. Here, we present the novel finding that DUOX2-generated ROS induce AEC death, leading to hyperoxia-induced lung injury.
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Supelco
Hydrogen peroxide solution, for ultratrace analysis, 30 % (w/w)
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
MISSION® esiRNA, targeting human NOX1
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fluorescein, free acid