MilliporeSigma
  • Evidence of the survival of ectopically transplanted oral mucosal epithelial stem cells after repeated wounding of cornea.
Evidence of the survival of ectopically transplanted oral mucosal epithelial stem cells after repeated wounding of cornea.
Molecular therapy : the journal of the American Society of Gene Therapy (2014-04-29)
Hiroaki Sugiyama, Masayuki Yamato, Kohji Nishida, Teruo Okano
ABSTRACT
Tissue engineering has become an essential tool in the development of regenerative medicine. We have developed cell sheet-based techniques for use in regenerative medicine that have already been successfully used in clinical applications. Native corneal epithelium is produced from limbal stem cells located in the transition zone between the cornea and the bulbar conjunctiva. Limbal stem cell deficiency (LSCD) is a severe defect of the limbal stem cells leading to vision loss due to conjunctival epithelial invasion and neovascularization. Rabbit LSCD models were treated with transplantable autologous oral mucosal epithelial cell (OEC) sheets fabricated on temperature-responsive cell culture surfaces, after which, the ocular surfaces were clear and smooth with no observable defects. The central part of the reconstructed ocular surface was scraped and wounded, after which proliferating epithelial cells covered the scraped area within a few days. The ocular surfaces were clear and smooth even after repeated scrapings and consisted of only OECs or heterogeneously mixed with corneal epithelial cells. This study demonstrates that transplanted cell sheets containing oral mucosal epithelial stem cells could reconstruct the ocular surface to maintain cornea homeostasis; moreover, they provide an ideal microenvironment to support the proliferation of remaining native limbal stem cells.
Product Description
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Betamethasone, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Betamethasone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Betamethasone, ≥98%
Supelco
Betamethasone, VETRANAL®, analytical standard
Supelco
Betamethasone, Pharmaceutical Secondary Standard; Certified Reference Material