MilliporeSigma
  • Cytotoxic activity of the amphibian ribonucleases onconase and r-amphinase on tumor cells from B cell lymphoproliferative disorders.
Cytotoxic activity of the amphibian ribonucleases onconase and r-amphinase on tumor cells from B cell lymphoproliferative disorders.
International journal of oncology (2014-05-03)
Piotr Smolewski, Magdalena Witkowska, Malgorzata Zwolinska, Barbara Cebula-Obrzut, Agata Majchrzak, Aleksandra Jeske, Zbigniew Darzynkiewicz, Wojciech Ardelt, Barbara Ardelt, Tadeusz Robak
ABSTRACT
Although major advancements in antitumor treatment have been observed, several B cell-derived malignancies still remain incurable. A promising approach that involves targeting RNA either by the use of specific antisense oligonucleotides or cytostatic/cytotoxic ribonucleases (RNases) is being promoted. Two amphibian RNases, onconase (ONC; ranpirnase) and, more recently, r-amphinase (r-Amph), have already been tested, but thus far, mostly on solid tumors. In this study, for the first time we provide comprehensive data on ex vivo and in vivo cytotoxic activity of ONC or r-Amph against cancer cells from different B cell lymphoid malignancies, together with their detailed mode of antitumor action. Our data revealed strong pro-apoptotic activity of ONC and r-Amph in both chronic lymphocytic leukemia and aggressive B cell lymphomas, with less impact on acute lymphoblastic leukemia or multiple myeloma cells. Moreover, the antitumor action of ONC and r-Amph was markedly selective against neoplastic cells sparing normal, healthy control‑derived lymphocytes.
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O