MilliporeSigma
  • Multifaceted roles of BDNF and FGF2 in human striatal primordium development. An in vitro study.
Multifaceted roles of BDNF and FGF2 in human striatal primordium development. An in vitro study.
Experimental neurology (2014-05-06)
Erica Sarchielli, Mirca Marini, Stefano Ambrosini, Alessandro Peri, Benedetta Mazzanti, Pamela Pinzani, Emanuela Barletta, Lara Ballerini, Ferdinando Paternostro, Marco Paganini, Berardino Porfirio, Annamaria Morelli, Pasquale Gallina, Gabriella B Vannelli
ABSTRACT
Grafting fetal striatal cells into the brain of Huntington's disease (HD) patients has raised certain expectations in the past decade as an effective cell-based-therapy for this devastating condition. We argue that the first requirement for successful transplantation is defining the window of plasticity for the striatum during development when the progenitor cells, isolated from their environment, are able to maintain regional-specific-identity and to respond appropriately to cues. The primary cell culture from human fetal striatal primordium described here consists of a mixed population of neural stem cells, neuronal-restricted progenitors and striatal neurons. These cells express trophic factors, such as BDNF and FGF2. We show that these neurotrophins maintain cell plasticity, inducing the expression of neuronal precursor markers and cell adhesion molecules, as well as promoting neurogenesis, migration and survival. We propose that BDNF and FGF2 play an important autocrine-paracrine role during early striatum development in vivo and that their release by fetal striatal grafts may be relevant in the setting of HD cell therapy.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Tyrosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Alizarin, Dye content 97 %
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
DL-Tyrosine, 99%
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
USP
Liothyronine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Indomethacin, European Pharmacopoeia (EP) Reference Standard
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine, ≥95% (HPLC), powder
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)