MilliporeSigma
  • Toll-like receptor 4 activation in Barrett's esophagus results in a strong increase in COX-2 expression.
Toll-like receptor 4 activation in Barrett's esophagus results in a strong increase in COX-2 expression.
Journal of gastroenterology (2013-08-21)
Romy E Verbeek, Peter D Siersema, Fiebo J Ten Kate, Kees Fluiter, Rhonda F Souza, Frank P Vleggaar, Pauline Bus, Jantine W P M van Baal
ABSTRACT
Barrett's esophagus (BE) is known to progress to esophageal adenocarcinoma in a setting of chronic inflammation. Toll-like receptor (TLR) 4 has been linked to inflammation-associated carcinogenesis. We aimed to determine the expression and functional activity of TLR4 in the esophagus and whether TLR4 activation in BE could promote carcinogenesis by inducing COX-2 expression. TLR4 expression in esophageal adenocarcinoma, BE, duodenum, reflux esophagitis and normal squamous esophagus biopsies was assessed using real-time PCR and validated by in situ hybridization and immunohistochemistry. Ex vivo cultures of BE, duodenum and normal squamous esophagus biopsies and a BE cell line (BAR-T) were stimulated with the TLR4 agonist lipopolysaccharide (LPS). To evaluate the effect of TLR4 activation, NF-κB activation, IL8 secretion and expression and COX-2 expression were determined. TLR4 expression was significantly increased in esophageal adenocarcinoma, BE, duodenum and reflux esophagitis compared to normal squamous esophagus. LPS stimulation resulted in NF-κB activation and a dose-dependent increase of IL8 secretion and mRNA expression. The induction of IL8 was more evident in BE compared to normal squamous esophagus. Upon LPS stimulation, COX-2 expression increased significantly in ex vivo cultured BE biopsies, which was observed in both epithelium and lamina propria cells. However, no effect was found in duodenum and normal squamous esophagus biopsies. TLR4 activation in BE results in a strong increase in COX-2 and may contribute to malignant transformation.
Product Description
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Tumor Necrosis Factor-α human, TNF-α, recombinant, expressed in E. coli, powder, suitable for cell culture
Sigma-Aldrich
Tumor Necrosis Factor-α human, Xeno-free, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
TNF-α human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human TLR4