MilliporeSigma
  • Platelet-derived nerve growth factor supports the survival of cholinergic neurons in organotypic rat brain slices.
Platelet-derived nerve growth factor supports the survival of cholinergic neurons in organotypic rat brain slices.
Neuroscience letters (2014-05-28)
Kathrin M Kniewallner, Natalia Grimm, Christian Humpel
ABSTRACT
Platelets play a role in repair of vessels and contain different growth factors, including nerve growth factor (NGF). Since NGF is the most potent growth factor to support survival of cholinergic neurons, we aimed to study the effects of platelet-derived NGF on cholinergic neurons in organotypic brain slices. Brain slices of the nucleus basalis of Meynert (nBM) were cultured with or without NGF (10ng/ml) or platelet extracts (100μg/ml) or fresh platelets (10(8) platelets/ml). In order to enhance NGF in platelets recombinant NGF (100ng) was loaded into platelets using ultrasound (3h). Our data show that recombinant NGF markedly supports survival of cholinergic neurons. The addition of fresh platelets showed a tendency for enhancing cholinergic neuron numbers, while platelet extracts had no effects. Ultrasound was highly effective to load recombinant NGF into platelets. The addition of NGF-loaded platelets markedly enhanced cholinergic neuron numbers. In conclusion, our data provide evidence that NGF-derived platelets may counteract cell death of cholinergic neurons.
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
SAFC
HEPES
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Methanol
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, Absolute - Acetone free
SAFC
HEPES
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%