MilliporeSigma
  • Intracellular alpha-synuclein affects early maturation of primary oligodendrocyte progenitor cells.
Intracellular alpha-synuclein affects early maturation of primary oligodendrocyte progenitor cells.
Molecular and cellular neurosciences (2014-07-16)
Benjamin Ettle, Simone Reiprich, Janina Deusser, Johannes C M Schlachetzki, Wei Xiang, Iryna Prots, Eliezer Masliah, Beate Winner, Michael Wegner, Jürgen Winkler
ABSTRACT
Myelin loss is a widespread neuropathological hallmark of the atypical parkinsonian disorder multiple system atrophy (MSA). On a cellular level, MSA is characterized by alpha-synuclein (aSyn)-positive glial cytoplasmic inclusions (GCIs) within mature oligodendrocytes leading to demyelination as well as axonal and neuronal loss. Oligodendrocyte progenitor cells (OPCs) represent a proliferative cell population distributed throughout the adult mammalian central nervous system. During remyelination, OPCs are recruited to sites of demyelination, differentiate, and finally replace dysfunctional mature oligodendrocytes. However, comprehensive studies investigating OPCs and remyelination processes in MSA are lacking. In the present study, we therefore investigate the effect of human aSyn (h-aSyn) on early primary rat OPC maturation. Upon lentiviral transduction, h-aSyn expressing OPCs exhibit fewer and shorter primary processes at the initiation of differentiation. Until day 4 of a 6day differentiation paradigm, h-aSyn expressing OPCs further show a severely delayed maturation evidenced by reduced myelin gene expression and increased levels of the progenitor marker platelet derived growth factor receptor-alpha (PDGFRα). Matching these results, OPCs that take up extracellular recombinant h-aSyn exhibit a similar delayed differentiation. In both experimental setups however, myelin gene expression is restored at day 6 of differentiation paralleled by decreased intracellular h-aSyn levels indicating a reverse correlation of h-aSyn and the differentiation potential of OPCs. Taken together, these findings suggest a tight link between the intracellular level of h-aSyn and maturation capacity of primary OPCs.
Sigma-Aldrich
Anti-Vimentin Antibody, clone V9, clone V9, Chemicon®, from mouse
Sigma-Aldrich
Anti-Olig2 Antibody, clone 211F1.1, clone 211F1.1, from mouse
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
DAPI, for nucleic acid staining
SAFC
BIS-TRIS
SAFC
BIS-TRIS
Sigma-Aldrich
Anti-CNPase Antibody, clone 11-5B, clone 11-5B, Chemicon®, from mouse
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)