MilliporeSigma
  • Serotonin 2A receptor regulates microtubule assembly and induces dynamics of dendritic growth cones in rat cortical neurons in vitro.
Serotonin 2A receptor regulates microtubule assembly and induces dynamics of dendritic growth cones in rat cortical neurons in vitro.
Neuroscience research (2014-04-05)
Akiko Ohtani, Naoki Kozono, Kouji Senzaki, Takashi Shiga
ABSTRACT
Serotonin (5-HT) regulates the development of cerebral cortex, but 5-HT receptors mediating the effects are poorly understood. We investigated roles of 5-HT2A receptor in dendritic growth cones using dissociation culture of rat cerebral cortex. Neurons at embryonic day 16 were cultured for 4 days and treated with 5-HT2A/2C receptor agonist (DOI) for 4h. DOI increased the size of growth cone periphery which was actin-rich and microtubule-associated protein 2-negative at the dendritic tip. The length increase of the growth cone periphery may be mediated by 5-HT2A receptor, because the 5-HT2A receptor antagonist reversed the effects of DOI. Moreover, the time-lapse analysis demonstrated the increase of morphological dynamics in dendritic growth cones by DOI. Next, to elucidate the mechanisms underlying the actions of 5-HT2A receptor in dendritic growth cones, we examined the cytoskeletal proteins, tyrosinated α-tubulin (Tyr-T; dynamic tubulin) and acetylated α-tubulin (Ace-T; stable tubulin). DOI increased the fluorescence intensity of Tyr-T, while decreased that of Ace-T in the dendritic growth cone periphery. These effects were reversed by the 5-HT2A receptor antagonist, suggesting that 5-HT2A receptor promotes microtubule dynamics. In summary, it was suggested that 5-HT2A receptor induces morphological changes and dynamics of dendritic growth cones through regulation of microtubule assembly.
Product Description
Cytarabine, European Pharmacopoeia (EP) Reference Standard
USP
Cytarabine, United States Pharmacopeia (USP) Reference Standard
Isoflurane, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source