MilliporeSigma
  • Microparticles released by vascular endothelial cells increase hypoxia inducible factor expression in human proximal tubular HK-2 cells.
Microparticles released by vascular endothelial cells increase hypoxia inducible factor expression in human proximal tubular HK-2 cells.
The international journal of biochemistry & cell biology (2014-06-01)
Ana Belen Fernandez-Martínez, Ana Valdehita Torija, Julia Carracedo, Rafael Ramirez, Francisco Javier de Lucio-Cazaña
ABSTRACT
Microparticles are produced by vesiculation of the cell plasma membrane and serve as vectors of cell-to-cell communication. Co-culture experiments have shown that hypoxia-inducible factor-α (HIF-α)-regulated-genes are up-regulated in human renal proximal tubular HK-2 cells by endothelial cell factors which might be transported inside endothelial microparticles (EMP). Here we aimed to study in HK-2 cells the effect of EMP, produced by activated endothelial cells, on HIF-α and HIF-α-regulated vascular endothelial growth factor-A (VEGF-A). EMP, at a concentration much lower than that found in plasma, increased the expression of HIF-α/VEGF-A in a COX-2/EP2 receptor dependent manner. Since the EMP/cells ratio was ∼1/1000, we hypothesized that paracrine mediators produced by HK-2 cells amplified the initial signal. This hypothesis was confirmed by two facts which also suggested that the mediators were conveyed by particles released by HK-2 cells: (i) HIF-α was up-regulated in HK-2 cells treated with the pellet obtained from the conditioned medium of the EMP-treated HK-2 cells. (ii) In transwell experiments, EMP-treated cells increased the expression of HIF-α in untreated HK-2 cells. Interestingly, we detected these cells, particles that were released by EMP-treated HK-2 cells. Depending on the pathological context, activation of HIF-α and VEGF-A signaling in renal tissue/cells may have either beneficial or harmful effects. Therefore, our results suggest that their presence in the urinary space of EMP produced by activated endothelial cells may influence the outcome of a number of renal diseases.
Product Description
Selenium, pellets, < 5mm, ≥99.999%
Celecoxib, European Pharmacopoeia (EP) Reference Standard
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis