MilliporeSigma
  • Insulin sensitivity is independent of lipid binding protein trafficking at the plasma membrane in human skeletal muscle: effect of a 3-day, high-fat diet.
Insulin sensitivity is independent of lipid binding protein trafficking at the plasma membrane in human skeletal muscle: effect of a 3-day, high-fat diet.
American journal of physiology. Regulatory, integrative and comparative physiology (2014-08-29)
Andreas B Jordy, Annette K Serup, Kristian Karstoft, Henriette Pilegaard, Bente Kiens, Jacob Jeppesen
ABSTRACT
The aim of the present study was to investigate lipid-induced regulation of lipid binding proteins in human skeletal muscle and the impact hereof on insulin sensitivity. Eleven healthy male subjects underwent a 3-day hypercaloric and high-fat diet regime. Muscle biopsies were taken before and after the diet intervention, and giant sarcolemmal vesicles were prepared. The high-fat diet induced decreased insulin sensitivity, but this was not associated with a relocation of FAT/CD36 or FABPpm protein to the sarcolemma. However, FAT/CD36 and FABPpm mRNA, but not the proteins, were upregulated by increased fatty acid availability. This suggests a time dependency in the upregulation of FAT/CD36 and FABPpm protein during high availability of plasma fatty acids. Furthermore, we did not detect FATP1 and FATP4 protein in giant sarcolemmal vesicles obtained from human skeletal muscle. In conclusion, this study shows that a short-term lipid-load increases mRNA content of key lipid handling proteins in human muscle. However, decreased insulin sensitivity after a high-fat diet is not accompanied with relocation of FAT/CD36 or FABPpm protein to the sarcolemma. Finally, FATP1 and FATP4 protein was located intracellularly but not at the sarcolemma in humans.
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioUltra, Molecular Biology, ≥99.5% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)