MilliporeSigma
  • Detection and characterisation of disseminated tumour cells in bone marrow of breast cancer patients by immunostaining of Her-2 and MUC-1 in combination with Thomsen-Friedenreich (CD176).
Detection and characterisation of disseminated tumour cells in bone marrow of breast cancer patients by immunostaining of Her-2 and MUC-1 in combination with Thomsen-Friedenreich (CD176).
Histology and histopathology (2014-01-09)
U Andergassen, A C Kölbl, M Zebisch, S Heublein, S Hutter, M Ilmer, C Schindlbeck, K Friese, U Jeschke
ABSTRACT
Disseminated tumour cells (DTCs) in the bone marrow derive from many primary tumours, such as breast cancer. Their mere existence hints to present or future metastasis and implicates a worse prognosis for the patient. DTCs may possess different characteristics in comparison to the primary tumour due to events like Epithelial-Mesenchymal-Transition. Therefore, these cells might be able to survive chemotherapy and cause relapses of the disease at a later point. We aimed to detect and further characterise DTCs by an immunostaining approach with three different antigen markers (Her-2, MUC-1 and TF, also known as CD 176). For that reason, bone marrow of 41 breast cancer patients was obtained during surgery; DTCs were enriched by density gradient centrifugation and cytospins were prepared. After fixation, immunofluorescent double-stainings were carried out with antibodies against CD176 in combination with HER-2 or MUC-1. Cells co-expressing two antigens were found in all staining combinations (Her-2 and CD176: 46.14%; MUC-1 and CD176: 18.15% of all cases). Cells that stained for a single antigen only were also found (Her-2: 36.86%; MUC-1: 34.45%; CD176: 29.65% of all cases). Significant correlations between the stainings of all markers could be shown (p<0,001). In conclusion, Thomsen-Friedenreich Antigen (TF, CD176) is a promising marker in combination with the established marker Her-2 and other markers like MUC-1. These results may serve as a basis for future DTC detection routines and help to individualize medical treatment, reducing side effects and increasing the efficiency of the therapy.
Product Description
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%