MilliporeSigma
  • Cytokine regulation by epidermal growth factor receptor inhibitors and epidermal growth factor receptor inhibitor associated skin toxicity in cancer patients.
Cytokine regulation by epidermal growth factor receptor inhibitors and epidermal growth factor receptor inhibitor associated skin toxicity in cancer patients.
European journal of cancer (Oxford, England : 1990) (2014-05-27)
Tanusree Paul, Christian Schumann, Stefan Rüdiger, Stefan Boeck, Volker Heinemann, Volker Kächele, Michael Steffens, Catharina Scholl, Vivien Hichert, Thomas Seufferlein, Julia Carolin Stingl
ABSTRACT
Epidermal growth factor receptor inhibitor (EGFRI) induced skin toxicity has a prognostic value suggesting skin toxicity can be a useful surrogate marker for successful epidermal growth factor receptor (EGFR) inhibition, improved response and survival. But the pathophysiology of EGFRI induced skin toxicity remains elusive. However the involvement of immunological mechanisms has been speculated. This study investigates the possible underlying mechanism of EGFR inhibition associated cytokine production in keratinocytes as well as in patients after treatment with epidermal growth factor receptor inhibitors (EGFRIs). Normal human epidermal keratinocytes (NHEK) were incubated for 2 and 24h with different concentrations of EGFRI (erlotinib) for Western blot analysis and cytokine expression analysis, respectively. Inhibition of EGFR, extracellular-signal-regulated kinase 1/2 (Erk 1/2) and c-Jun was examined by Western blot analysis. Cytokine concentrations were measured by enzyme-linked immunosorbent assay (ELISA) in the NHEK cell supernatant and also in the serum of 186 cancer patients who are followed up for EGFRI induced skin rash. A significant inhibitory effect of EGFRI was seen on EGFR (Y845), Erk 1/2 and c-Jun in a dose dependent manner. Further downstream, increased CC-chemokine ligand 2 (CCL2), CC-chemokine ligand 5 (CCL5) and decreased interleukin-8 (IL-8) or CXCL8 expression was observed in keratinocytes. In EGFRI treated patients, low levels of serum CXCL8 corresponding to stronger EGFR inhibition were associated with a higher grade of skin toxicity (p=0.0016) and a prolonged overall survival (p=0.018). The results obtained in this study indicate that EGFRI can regulate cytokines by modulating EGFR signalling pathway in keratinocytes. Moreover, serum levels of CXCL8 in EGFRI treated patients may be important for individual EGFRI induced skin toxicity and patient's survival.
Product Description
SAFC
Sodium deoxycholate
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG