MilliporeSigma
  • Dual effects of amiodarone on pacemaker currents in hypertrophied ventricular myocytes isolated from spontaneously hypertensive rats.
Dual effects of amiodarone on pacemaker currents in hypertrophied ventricular myocytes isolated from spontaneously hypertensive rats.
Clinical and experimental pharmacology & physiology (2014-05-28)
Hongxia Li, Yafeng Zhou, Bin Jiang, Xin Zhao, Xun Li, Xiangjun Yang, Wenping Jiang
ABSTRACT
The pacemaker current If conducted by hyperpolarization-activated cyclic nucleotide-gated (HCN) channels plays a critical role in the regulation of cardiac automaticity, with If density increased in hypertrophied ventricular myocytes. Amiodarone, a highly effective anti-arrhythmic agent, blocks human HCN currents and native If under normal conditions. To determine the effects of amiodarone under pathological conditions, we monitored If under after both acute (0.01, 0.1, 1, 10 and 100 μmol/L) and chronic (10 μmol/L) amiodarone treatment in ventricular myocytes from spontaneously hypertensive rats (SHR) with left ventricular hypertrophy using the whole-cell patch-clamp technique. The If current density was significantly greater in SHR ventricular myocytes than in cells from healthy normotensive control Wistar-Kyoto (WKY) rats. Acute application of amiodarone significantly decreased If density in myocytes from both SHR and WKY rats. The inhibition was concentration dependent with an IC50 of 4.9 ± 1.2 and 6.9 ± 1.3 μmol/L in myocytes from SHR and WKY rats, respectively. Amiodarone increased the activation and deactivation times of If in myocytes from SHR, although it did not alter the relationship of voltage-dependent activation and the reversal potential of If in myocytes from SHR. Chronic exposure of myocytes from SHR to amiodarone potently inhibited If and downregulated HCN2 and HCN4, the major channel subtypes underlying native If , at both the mRNA and protein level. These findings indicate that amiodarone inhibits If under hypertrophied conditions through dual mechanisms: (i) direct channel blockade of If currents; and (ii) indirect suppression via negative regulation of HCN channel gene expression. These unique properties of amiodarone may contribute to its anti-arrhythmic properties under pathological conditions.
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, BioUltra, ~0.025 M in H2O
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 1.00 M±0.01 M
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
USP
Monobasic potassium phosphate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis