MilliporeSigma
  • Chronic toluene exposure induces cell proliferation in the mice SVZ but not migration through the RMS.
Chronic toluene exposure induces cell proliferation in the mice SVZ but not migration through the RMS.
Neuroscience letters (2014-06-03)
Ireri Franco, Mariana Valdez-Tapia, Sinthia L Sanchez-Serrano, Silvia L Cruz, Monica Lamas
ABSTRACT
Abuse of toluene-containing inhalants is associated to various cognitive impairments that have been partly associated to deviation of the hippocampal neurogenesis processes during adulthood. In the present study we analyzed the effect of chronic toluene exposure (6000ppm) on cell proliferation and migration in the other selected area of the rodent brain where neurogenesis persist throughout adulthood, the subventricular zone of the lateral ventricle (SVZ). We used an anti-Ki67 antibody to evaluate SVZ cell proliferation, BrdU to evaluate cell survival and double-staining with BrdU and the migration marker doublecortin (DCX) to evaluate migration, by immunofluorescence 2h, 1, 5, 10 or 15 days after 20 sessions of toluene exposure. We found that toluene induced an initial burst of cell proliferation in the SVZ but not a significant increase in migration toward the rostral migratory stream (RMS) or the number of cells that migrate to the olfactory bulb. In addition, we detected a small number of new migrating cells in the corpus callosum and striatum of control mice that was similar in toluene-exposed brains. These results may underline the homeostatic capabilities of the populations of dividing cells, previously demonstrated using other drugs of abuse and demonstrate that toluene misuse can alter cellular proliferation in the postnatal brain.
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Supelco
Toluene, analytical standard
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material