MilliporeSigma
  • Angiogenic/osteogenic response of BMMSCs on bone-derived scaffold: effect of hypoxia and role of PI3K/Akt-mediated VEGF-VEGFR pathway.
Angiogenic/osteogenic response of BMMSCs on bone-derived scaffold: effect of hypoxia and role of PI3K/Akt-mediated VEGF-VEGFR pathway.
Biotechnology journal (2014-01-15)
Yi Zhou, Xiaoxu Guan, Mengfei Yu, Xinhua Wang, Wenyuan Zhu, Chaowei Wang, Mengliu Yu, Huiming Wang
ABSTRACT
Bone tissue deficiency is a common clinical challenge. Tissue-engineered bone constructs are an effective approach for the repair of orthopedic bone defects. Mimicking the essential components of the in vivo microenvironment is an efficient way to develop functional constructs. In this study, bone marrow-derived mesenchymal stromal cells (BMMSCs) were seeded into bone-derived scaffolds, a material with similar structure to natural bone. This was done under hypoxic conditions, an environment that imitates that experienced by BMMSCs in vivo. Our data indicate that hypoxia (5% O2 ) significantly increases the proliferation of BMMSCs seeded in scaffolds. As reflected by highly expressed osteogenesis- and angiogenesis-associated biomarkers, including vascular endothelial growth factor (VEGF), RUNX2, bone morphogenetic protein-2/4 and osteopontin, hypoxia also significantly increases the osteogenic and angiogenic responses of BMMSCs seeded into bone-derived scaffold composites. PI3K/Akt-mediated regulation of VEGF-activated VEGFR1/2 signaling is important for hypoxia-induced proliferative/osteogenic/angiogenic responses in BMMSC cellular scaffolds. The combination of bone-derived scaffolds and hypoxia is conducive to the differentiation of BMMSCs into functional tissue-engineered scaffold composites.
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water