MilliporeSigma
  • RASopathy-associated CBL germline mutations cause aberrant ubiquitylation and trafficking of EGFR.
RASopathy-associated CBL germline mutations cause aberrant ubiquitylation and trafficking of EGFR.
Human mutation (2014-09-03)
Kristina Brand, Hendrik Kentsch, Christina Glashoff, Georg Rosenberger
ABSTRACT
Noonan syndrome, a congenital disorder comprising a characteristic face, short stature, heart defects, learning difficulties, and a predisposition to malignancies, is caused by heterozygous germline mutations in genes encoding components of RAS-MAPK signaling pathways. Mutations in the CBL tumor suppressor gene have been reported in patients with a Noonan syndrome-like phenotype. CBL encodes a multivalent adaptor protein with ubiquitin ligase activity, which promotes ubiquitylation and vesicle-mediated internalization and degradation of the epidermal growth factor (EGF) receptor (EGFR). We investigated the functional consequences of disease-associated CBL amino acid changes p.K382E, p.D390Y, and p.R420Q on ligand-induced EGFR trafficking. Expression of CBL(K382E), CBL(D390Y), or CBL(R420Q) in COS-7 cells resulted in increased levels of surface EGFR and reduced amounts of intracellular EGFR; both consequences indicate ineffective EGFR internalization. Accordingly, receptor-mediated uptake of EGF was decreased. Furthermore, the p.K382E, p.D390Y, and p.R420Q lesions impaired CBL-mediated EGFR ubiquitylation and degradation. Together, these data indicate that pathogenic CBL mutations severely affect vesicle-based EGFR trafficking. Since we detected enhanced ERK phosphorylation in cells expressing mutant CBL, we conclude that aberrant EGFR trafficking contributes to augmented RAS-MAPK signaling, the common trait of Noonan syndrome and related RASopathies. Thus, our data suggest that EGFR trafficking is a novel disease-relevant regulatory level in the RASopathy network.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
N-Ethylmaleimide, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, purum p.a., ≥99.0% (HPLC)
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, BioXtra, ≥98% (HPLC)