MilliporeSigma
  • Epstein-Barr virus encoded microRNAs target SUMO-regulated cellular functions.
Epstein-Barr virus encoded microRNAs target SUMO-regulated cellular functions.
The FEBS journal (2014-09-11)
Simone Callegari, Stefano Gastaldello, Omid R Faridani, Maria G Masucci
ABSTRACT
Post-translational modification by the small ubiquitin-like modifier (SUMO) regulates the cellular response to different types of stress and plays a pivotal role in the control of oncogenic viral infections. Here we investigated the capacity of microRNAs (miRNAs) encoded by Epstein-Barr virus to interfere with the SUMO signaling network. Using a computational strategy that scores different properties of miRNA-mRNA target pairs, we identified a minimal set of 575 members of the SUMO interactome that may be targeted by one or more Epstein-Barr virus miRNAs. A significant proportion of the candidates cluster in a functional network that controls chromatin organization, stress, DNA damage and immune responses, apoptosis and transforming growth factor beta signaling. Multiple components of the transforming growth factor beta signaling pathway were inhibited upon upregulation of the BamHI-H rightward open reading frame 1 (BHRF1) encoded miRNAs in cells transduced with recombinant lentiviruses or entering the productive virus cycle. These findings point to the capacity of viral miRNAs to interfere with SUMO-regulated cellular functions that control key aspects of viral replication and pathogenesis.
Product Description
Supelco
Acrylamide, analytical standard
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Anti-SMAD3 antibody produced in rabbit
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Acrylamide, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered