MilliporeSigma
  • Brexpiprazole II: antipsychotic-like and procognitive effects of a novel serotonin-dopamine activity modulator.
Brexpiprazole II: antipsychotic-like and procognitive effects of a novel serotonin-dopamine activity modulator.
The Journal of pharmacology and experimental therapeutics (2014-06-21)
Kenji Maeda, Linda Lerdrup, Haruhiko Sugino, Hitomi Akazawa, Naoki Amada, Robert D McQuade, Tine Bryan Stensbøl, Christoffer Bundgaard, Jørn Arnt, Tetsuro Kikuchi
ABSTRACT
Brexpiprazole (OPC-34712, 7-{4-[4-(1-benzothiophen-4-yl)piperazin-1-yl]butoxy}quinolin-2(1H)-one) is a novel serotonin-dopamine activity modulator with partial agonist activity at serotonin 1A (5-HT1A) and D2/3 receptors, combined with potent antagonist effects on 5-HT2A, α1B-, and α2C-adrenergic receptors. Brexpiprazole inhibited conditioned avoidance response (ED50 = 6.0 mg/kg), apomorphine- or d-amphetamine-induced hyperactivity (ED50 = 2.3 and 0.90, respectively), and apomorphine-induced stereotypy (ED50 = 2.9) in rats at clinically relevant D2 receptor occupancies. Brexpiprazole also potently inhibited apomorphine-induced eye blinking in monkeys. The results suggest that brexpiprazole has antipsychotic potential. Brexpiprazole induced catalepsy (ED50 = 20) well above clinically relevant D2 receptor occupancies, suggesting a low risk for extrapyramidal side effects. Subchronic treatment with phencyclidine (PCP) induced cognitive impairment in both novel object recognition (NOR) and attentional set-shifting (ID-ED) tests in rats. Brexpiprazole reversed the PCP-induced cognitive impairment in the NOR test at 1.0 and 3.0 mg/kg, and in the ID-ED test at 1.0 mg/kg. However, aripiprazole (10 mg/kg) was ineffective in both tests, despite achieving relevant D2 occupancies. In the NOR test, the 5-HT1A agonist buspirone and the 5-HT2A antagonist M100907 [(R)-(2,3-dimethoxyphenyl)[1-(4-fluorophenethyl)piperidin-4-yl]methanol] partially but significantly reversed PCP-induced impairment. Furthermore, the effect of brexpiprazole was reversed by cotreatment with the 5-HT1A antagonist WAY100635 (N-[2-[4-(2-methoxyphenyl)-1-piperazinyl]ethyl]-N-2-pyridinylcyclohexanecarboxamide maleate). The results indicate that brexpiprazole has antipsychotic-like activity and robust efficacy in relevant models of cognitive impairment associated with schizophrenia. The effects of brexpiprazole in the cognitive tests are superior to those of aripiprazole. We propose that the pharmacologic profile of brexpiprazole be based on its balanced effects on 5-HT1A, D2, and 5-HT2A receptors, with possible modulating activity through additional monoamine receptors.
Product Description
Risperidone for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Risperidone, United States Pharmacopeia (USP) Reference Standard
Aripiprazole, European Pharmacopoeia (EP) Reference Standard
Risperidone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Aripiprazole, ≥98% (HPLC)
Apomorphine hydrochloride hemihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
R-(−)-Apomorphine hydrochloride hemihydrate, calcined, ≥98% (with NaOH, titration)
Sigma-Aldrich
Risperidone, ≥98% (HPLC), powder