MilliporeSigma
  • FOXO3a potentiates hTERT gene expression by activating c-MYC and extends the replicative life-span of human fibroblast.
FOXO3a potentiates hTERT gene expression by activating c-MYC and extends the replicative life-span of human fibroblast.
PloS one (2014-07-08)
Shuntaro Yamashita, Kaori Ogawa, Takahiro Ikei, Tsukasa Fujiki, Yoshinori Katakura
ABSTRACT
In our previous studies, we reported that SIRT1 prevents cellular senescence in human fibroblast, and that SIRT1-induced inhibition of cellular senescence is due to enhanced hTERT gene expression. In this study, we investigate the molecular mechanisms behind SIRT1-induced potentiation of hTERT transcription and show that FOXO3a functions downstream of SIRT1 and prevents the induction of cellular senescence by enhancing hTERT gene expression. Furthermore, we found that FOXO3a-induced potentiation of hTERT gene expression is regulated in a c-MYC/E-box dependent manner. In addition, we found that FOXO3a binds to the novel binding element in the c-MYC promoter, and this interaction activates the transcription of the c-MYC gene. The resulting increase in c-MYC leads to higher levels of c-MYC recruited to the hTERT promoter and, in turn, activates hTERT gene expression. Taken together, this pathway might constitute the molecular basis for the anti-senescence effects of SIRT1 and FOXO3a.
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-c-Myc antibody produced in rabbit, ~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Anti-NOL3 antibody produced in rabbit, affinity isolated antibody