MilliporeSigma
  • Inhibition of non-small cell lung cancer (NSCLC) growth by a novel small molecular inhibitor of EGFR.
Inhibition of non-small cell lung cancer (NSCLC) growth by a novel small molecular inhibitor of EGFR.
Oncotarget (2015-03-03)
Jinsong Li, Huayun Deng, Meichun Hu, Yuanzhang Fang, Amanda Vaughn, Xiaopan Cai, Leqin Xu, Wei Wan, Zhenxi Li, Shijie Chen, Xinghai Yang, Song Wu, Jianru Xiao
ABSTRACT
The epidermal growth factor receptor (EGFR) is a therapeutic target (oncotarget) in NSCLC. Using in vitro EGFR kinase activity system, we identified a novel small molecule, WB-308, as an inhibitor of EGFR. WB-308 decreased NSCLC cell proliferation and colony formation, by causing G2/M arrest and apoptosis. Furthermore, WB-308 inhibited the engraft tumor growths in two animal models in vivo (lung orthotopic transplantation model and patient-derived engraft mouse model). WB-308 impaired the phosphorylation of EGFR, AKT, and ERK1/2 protein. WB-308 was less cytotoxic than Gefitinib. Our study suggests that WB-308 is a novel EGFR-TKI and may be considered to substitute for Gefitinib in clinical therapy for NSCLC.
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)