MilliporeSigma
  • Downregulated expression of PTK6 is correlated with poor survival in esophageal squamous cell carcinoma.
Downregulated expression of PTK6 is correlated with poor survival in esophageal squamous cell carcinoma.
Medical oncology (Northwood, London, England) (2014-11-08)
You-Fang Chen, Gang Ma, Xun Cao, Zhi-Liang Huang, Mu-Sheng Zeng, Zhe-Sheng Wen
ABSTRACT
To investigate the clinical prognostic value of protein tyrosine kinase 6 (PTK6) in patients with esophageal squamous cell carcinoma (ESCC), quantitative RT-PCR and Western blotting were utilized to measure the mRNA and protein expression levels of PTK6 in 29 and eight pairs of ESCC and peritumoral normal esophageal tissues, respectively. Furthermore, the expression of PTK6 protein in 210 ESCCs was examined with immunohistochemistry (IHC), and its clinical value was analyzed using Kaplan-Meier plots and the Cox proportional hazards regression model. The results found that the expression levels of both PTK6 mRNA and protein in ESCC tissues were significantly lower than those in peritumoral normal esophageal tissues. Regarding the IHC analysis of ESCC, the cytoplasmic expression of PTK6 was significantly correlated with tumor grade (P < 0.001). Compared with patients with low PTK6 expression, ESCC patients with overexpression of PTK6 displayed preferable disease-free survival (DFS) and overall survival (OS) (P < 0.001 and P = 0.001, respectively), especially in stage II disease (P = 0.002 and P = 0.021, respectively). PTK6 was evaluated as an independent prognostic factor for ESCC using multivariate Cox regression analysis. All data demonstrated that the expression level of PTK6 is an independent prognostic factor in ESCCs. Low expression of PTK6 is correlated with poor DFS and OS in ESCCs.
Product Description
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Supelco
Glycerol, analytical standard
Supelco
2-Mercaptoethanol, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))