MilliporeSigma
  • Processing bodies accumulate in human cytomegalovirus-infected cells and do not affect viral replication at high multiplicity of infection.
Processing bodies accumulate in human cytomegalovirus-infected cells and do not affect viral replication at high multiplicity of infection.
Virology (2014-06-15)
Eri Seto, Teruki Inoue, Yoko Nakatani, Masanobu Yamada, Hiroki Isomura
ABSTRACT
Translationally silenced mRNAs are recruited to two major classes of RNA granules in the cytoplasm, processing bodies (PBs) and stress granules (SGs). We show that PBs accumulated after human cytomegalovirus (HCMV) infection. PB assembly after HCMV infection was also detected in the presence of the protein synthesis inhibitor, cycloheximide, but required active RNA synthesis. UV-inactivated HCMV virions were sufficient to induce PB accumulation in HFF cells treated with cycloheximide. Viral IE1 RNA did not colocalize with PBs, and we could not detect an effect of PB accumulation on viral growth. These results may indicate that HCMV inhibits the colocalization of IE1 mRNA with PBs, preventing IE1 mRNA decay and translational inhibition.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining