MilliporeSigma
  • Role of the Wilms' tumor 1 gene in the aberrant biological behavior of leukemic cells and the related mechanisms.
Role of the Wilms' tumor 1 gene in the aberrant biological behavior of leukemic cells and the related mechanisms.
Oncology reports (2014-10-14)
Yan Li, Jiying Wang, Xiaoyan Li, Yujiao Jia, Lei Huai, Kan He, Pei Yu, Min Wang, Haiyan Xing, Qing Rao, Zhen Tian, Kejing Tang, Jianxiang Wang, Yingchang Mi
ABSTRACT
The Wilms' tumor 1 (WT1) gene is one of the regulating factors in cell proliferation and development. It is a double-functional gene: an oncogene and a tumor suppressor. This gene was found to be highly expressed in many leukemic cell lines and in patients with acute myeloid leukemia. In the present study, we demonstrated that the WT1 gene was commonly expressed in leukemic cell lines apart from U937 cells. The K562 cell line which expresses WT1 at a high level (mRNA and protein) was used in the entire experiment. By MTT and colony formation assays, we found that curcumin, an inhibitor of the WT1 protein, inhibited cell proliferation and clonogenicity in a time- and dose-dependent manner. It also caused cell cycle arrest at the G2/M phase. We then designed specific short hairpin RNAs (shRNAs) which could downregulate WT1 by 70-80% at the mRNA and protein levels. Reduction in the WT1 levels attenuated the proliferative ability and clonogenicity. Cell cycle progression analysis indicated that the proportion of cells in the G0/G1 phase increased while the proportion in the S phase decreased distinctively. ChIP-DNA selection and ligation (DSL) experiment identified a cohort of genes whose promoters are targeted by WT1. These genes were classified into different cellular signaling pathways using MAS software and included the Wnt/β-catenin pathway, MAPK signaling pathway, apoptosis pathway, and the cell cycle. We focused on the Wnt/β-catenin signaling pathway, and compared expression of several genes in the K562 cells transfected with the control shRNA and WT1-specific shRNA. β-catenin, an important gene in the Wnt canonical pathway, was downregulated after WT1 RNAi. Target genes of β-catenin which participate in cell proliferation and cell cycle regulation, such as CCND1 and MYC, were also significantly downregulated. Collectively, these data suggest that WT1 functions as an oncogene in leukemia cells, and one important mechanism is regulation of the Wnt/β-catenin pathway.
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma