MilliporeSigma
  • New proteomic insights on the role of NPR-A in regulating self-renewal of embryonic stem cells.
New proteomic insights on the role of NPR-A in regulating self-renewal of embryonic stem cells.
Stem cell reviews (2014-05-07)
Sameh Magdeldin, Tadashi Yamamoto, Ikuo Tooyama, Essam M Abdelalim
ABSTRACT
Embryonic stem cells (ESCs) have the ability to self-renew indefinitely and they can give unlimited source of cells and tissues for cellular therapies. Recently, the natriuretic peptide receptor A (NPR-A) has been recognized as an important regulator for the self-renewal of ESCs. To gain insights into possible novel mechanisms involved in NPR-A pathway that presumably regulates self-renewal and survival of ESCs, we utilized a comprehensive label-free proteomics technology in our study. Targeting of NPR-A gene with small interfering RNA (siRNA) resulted in the inhibition of ESCs self-renewal. Coherently, quantitative label-free shotgun proteomic analysis identified differentially expressed proteins involved in several biological processes, including cell cycle regulation, cell proliferation, cell fate specification, and apoptosis. Interestingly, in addition to Oct4 Nanog, and Sox2, other proteins involved in ESCs self-renewal were down-regulated after NPR-A knockdown, such as heterogeneous nuclear ribonucleoprotein A2/B1 (ROA2), non-POU domain-containing octamer-binding protein (Nono), nucleoplasmin (Npm1), histone H2A type 1-B/E (histone H2A.2), SW1/SNF complex (Brg1), polycomb protein Suz12 (Suz12), and cyclin-dependent kinase 4 (Cdk4). Furthermore, several protein candidates involved in early differentiation and cell death were up-regulated or down-regulated as a result of NPR-A knockdown, including importin subunit alpha-4 (Impα4), importin-5 (Ipo5), H3 histones, core histone macro-H2A.1 (H2A.y), apurine/apyrimidine endonuclease 1 (Apex1), 78-kDa glucose-regulated protein (Grp78), and programmed cell death 5 (Pdcd5). Overall, these findings depict a comprehensive view to our understanding of the pathways involved in the role of NPR-A in maintaining ESC functions.
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Supelco
2-Mercaptoethanol, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%