MilliporeSigma
  • MiR-132 inhibits expression of SIRT1 and induces pro-inflammatory processes of vascular endothelial inflammation through blockade of the SREBP-1c metabolic pathway.
MiR-132 inhibits expression of SIRT1 and induces pro-inflammatory processes of vascular endothelial inflammation through blockade of the SREBP-1c metabolic pathway.
Cardiovascular drugs and therapy (2014-06-14)
Liwei Zhang, Dangsheng Huang, Qiushuang Wang, Dong Shen, Yumei Wang, Bingyang Chen, Jinqian Zhang, Luyue Gai
ABSTRACT
Inflammation participates centrally in all stages of atherosclerosis (AS), which begins with pro-inflammatory processes and inflammatory changes in the endothelium, related to lipid metabolism. MicroRNA (miRNA) inhibition of inflammation related to SIRT1 has been shown to be a promising therapeutic approach for AS. However, the mechanism of action is unknown. We investigated whether miRNAs regulate the SIRT1 and its downstream SREBP-lipogenesis-cholesterogenesis metabolic pathway in human umbilical vein endothelial cells (HUVECs). HUVECs were transfected with miR-132 mimics and inhibitors, and then treated with or without tumor necrosis factor α (TNFα). The effects of miR-132 on pro-inflammatory processes, proliferation and apoptosis were assessed. We identified that the relative 3' UTR luciferase activities of SIRT1 were significantly decreased in miR-132 transfected HUVECs (0.338 ± 0.036) compared to control (P = 0.000). miR-132 inhibited SIRT1 expression of mRNA level in HUVECs (0.53 ± 0.06) (P < 0.01) as well as proteins of SIRT1. mRNA expression and protein levels of SREBP (0.45 ± 0.07), fatty acid synthase (FASN) (0.55 ± 0.09) and 3-hydroxy-3-methylglutaryl CoA reductase (HMGCR) (0.62 ± 0.08) (P < 0.01), which are downstream regulated genes, were reduced in HUVECs by miR-132. MiR-132 promoted pro-inflammatory processes and apoptosis of HUVECs induced by TNF-α, and inhibited its proliferation, viability and migration. SIRT1 mRNAs are direct targets of miR-132. miR-132 controls lipogenesis and cholesterogenesis in HUVECs by inhibiting SIRT1 and SREBP-1c expression and their downstream regulated genes, including FASN and HMGCR. Inhibition of SIRT1 by miR-132 was associated with lipid metabolism-dependent pro-inflammatory processes in HUVECs. The newly identified miRNA, miR-132 represents a novel targeting mechanism for AS therapy.
Product Description
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterol, Plant-Derived, SyntheChol®