MilliporeSigma
Anti-amyloidogenic property of human gastrokine 1.
Biochimie (2014-08-21)
Filomena Altieri, Chiara Stella Di Stadio, Valeria Severino, Annamaria Sandomenico, Giuseppina Minopoli, Giuseppina Miselli, Antimo Di Maro, Menotti Ruvo, Angela Chambery, Vincenzo Quagliariello, Mariorosario Masullo, Emilia Rippa, Paolo Arcari
ABSTRACT
Gastrokine 1 (GKN1) is a stomach-specific protein expressed in normal gastric tissue but absent in gastric cancer. GKN1 plays a major role in maintaining gastric mucosa integrity and is characterized by the presence of a BRICHOS domain consisting of about 100 amino acids also found in several unrelated proteins associated with major human diseases like BRI2, related to familial British and Danish dementia and surfactant protein C (SP-C), associated with respiratory distress syndrome. It was reported that recombinant BRICHOS domains from BRI2 and SP-C precursor (proSP-C) prevent fibrils formation of amyloid-beta peptide (Aβ), that is the major component of extracellular amyloid deposits in Alzheimer's disease. Here we investigated on the interaction between human recombinant GKN1 (rGKN1) and Aβ peptide (1-40) that derives from the partial hydrolysis of the amyloid precursor protein (APP). GKN1 prevented amyloid aggregation and fibrils formation by inhibiting Aβ(1-40) polymerization, as evaluated by SDS-PAGE, thioflavin-T binding assay and gel filtration experiments. Mass spectrometry showed the formation of a prevailing 1:1 complex between GKN1 and Aβ(1-40). SPR analysis of GKN1/Aβ interaction led to calculate a dissociation constant (KD) of 34 μM. Besides its interaction with Aβ(1-40), GKN1 showed also to interact with APP as evaluated by confocal microscopy and Ni-NTA pull-down. Data strongly suggest that GKN1 has anti-amyloidogenic properties thus functioning as a chaperone directed against unfolded segments and with the ability to recognize amyloidogenic polypeptides and prevent their aggregation.
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O