MilliporeSigma
  • Keratocytes derived from spheroid culture of corneal stromal cells resemble tissue resident keratocytes.
Keratocytes derived from spheroid culture of corneal stromal cells resemble tissue resident keratocytes.
PloS one (2014-11-11)
Yong-Soo Byun, Sapna Tibrewal, Eunjae Kim, Lisette Yco, Joy Sarkar, Yair Ivanir, Chia-Yang Liu, Cecile M Sano, Sandeep Jain
ABSTRACT
Corneal stromal cells transform to precursor cells in spheroid culture. We determined whether keratocytes derived from spheroid culture of murine corneal stromal cells resemble tissue resident keratocytes. Spheroid culture was performed by seeding dissociated stromal cells onto ultra-low attachment plates containing serum-free mesenchymal stem cell culture medium. Spheroids were characterized with phenotype specific markers and stemness transcription factor genes. Spheroids and adherent cells in culture were induced to differentiate to keratocytes using keratocyte induction medium (KIM) and compared with tissue resident keratocytes. Stromal cells formed spheroids in ultra-low attachment plates, but not in polystyrene tissue culture dishes. Keratocan expression and abundance was significantly higher in spheroids as compared to adherent cells whereas alpha-smooth muscle actin (α-SMA) was significantly lower. As compared to adherent culture-derived cells, the expressions of keratocan, aldehyde dehydrogenase (ALDH3A1) and α-SMA in spheroid-derived cells approximated much more closely the levels of these genes in tissue resident keratocytes. Of the stemness genes, Nanog and Oct4 were upregulated in the spheroids. Stemness transcription factor genes are upregulated in spheroids. Keratocytes derived from spheroids resemble tissue resident keratocytes, thus increasing manifolds the quantity of these cells for in-vitro experiments.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
FGF-2 human, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
Sigma-Aldrich
FGF-2 human, recombinant, expressed in insect cells, ≥85% (SDS-PAGE)
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Calcein-AM, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)