MilliporeSigma
Chemical modification of hyaluronic acid for intraoral application.
Journal of pharmaceutical sciences (2014-07-06)
Flavia Laffleur, Julia Röggla, Muneeb Ahmad Idrees, Julia Griessinger
ABSTRACT
This study was aimed to investigate chemical preactivated thiomers for their potential use in mucosal drug delivery. Thiomers--thiolated polymers--are mucoadhesive polymers with sulfhydryl group-bearing side chains. Thiomers are synthesized by covalent attachment of low molecular mass compounds bearing sulfhydryl group to the polymeric backbone of well-established polymers. Hyaluronic acid-cysteine ethyl ester-mercaptonicotinamide conjugates (HA-CYS-MNA) were synthesized by the oxidative S-S coupling of HA-CYS with 6-MNA. Conjugates were compressed into test discs to investigate cohesive properties, cytotoxicity assays, and mucoadhesion studies. Because of the immobilization of MNA, the HA-CYS-MNA conjugates exhibit comparatively higher swelling properties and cohesive properties corresponding unmodified HA. On the rotating cylinder, discs based on HA-CYS-MNA conjugates displayed fourfold improved mucoadhesion time compared with thiolated polymers. Tensile study results were found in good agreement with rotating cylinder results. Moreover, preactivated thiomers showed higher stability. All polymers were found nontoxic over Caco-2 cells. On the basis of achieved results, the preactivated thiomeric therapeutic agent seems to represent a promising generation of mucoadhesive polymers that are safe to use for a prolonged residence time to target the mucosa requested for intraoral application.
Product Description
Supelco
Hydrogen peroxide solution, for ultratrace analysis, 30 % (w/w)
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
6-Chloronicotinamide, 98%
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Sigma-Aldrich
L-Cysteine ethyl ester hydrochloride, 98%