MilliporeSigma
  • Structural, optical and sensing properties of novel Eu(III) complexes with furan- and pyridine-based ligands.
Structural, optical and sensing properties of novel Eu(III) complexes with furan- and pyridine-based ligands.
Dalton transactions (Cambridge, England : 2003) (2014-11-06)
Fabio Piccinelli, Marco Bettinelli, Andrea Melchior, Cristian Grazioli, Marilena Tolazzi
ABSTRACT
A new family of imine and amine-based racemic ligands containing furan or pyridine as an aromatic donating ring [N,N′-bis(2-pyridylmethylidene)-1,2-(R,R + S,S)-cyclohexanediamine, L1; N,N′-bis(2-furanylmethylidene)-1,2-(R,R + S,S)-cyclohexanediamine, L2; N,N′-bis(2-pyridylmethyl)-1,2-(R,R + S,S)-cyclohexanediamine, L3; and N,N′-bis(2-furanylmethyl)-1,2-(R,R + S,S)-cyclohexanediamine, L4] and their trifluoromethanesulphonate (CF3SO3(−), OTf(−)) and nitrate Eu(III) complexes is studied in acetonitrile (AN) solution. The stoichiometry and stabilities of the formed complexes are obtained by means of spectrophotometric titrations: when Eu(III) triflate is used as a starting salt, two mononuclear species (1:1 and 1:2) are detected, while only the 1:1 complex is observed when the nitrate salt is employed. The stability of these complexes, as well as the geometry of their Eu(III) environment, is significantly dependent on the nature of the ligand employed (imine or amine, furan or pyridine-based). DFT calculations show that all donor atoms are coordinated to the metal ion in the 1:1 EuL(L = L1–L4) species and suggest that the higher stability of the complexes with L1 and L2 with respect to L3 and L4 is mostly due to the higher degree of preorganization of the former species. The optical response of the imine-based L1 and L2 Eu complexes, produced by NO3(−) coordination, has been studied in order to assess their application as sensing devices. With both ligands, an increase of the emission intensity on the addition of the nitrate ion is observed. This is higher for the EuL2 complex and underlines the important role of the nature of the heteroaromatic ring. Finally, it is worth noting that an efficient energy transfer process from the ligand to the metal is present in the case of the 1:1 triflate Eu(III) complex with the ligand L1.
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetonitrile, analytical standard
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Tetraethylammonium nitrate, ≥98.0% (NT)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material