MilliporeSigma
  • Glucocorticoid receptor function is decreased in neutrophils during endotoxic shock.
Glucocorticoid receptor function is decreased in neutrophils during endotoxic shock.
The Journal of infection (2014-03-25)
Maria Bergquist, Pernilla Jirholt, Merja Nurkkala, Christian Rylander, Göran Hedenstierna, Catharina Lindholm
ABSTRACT
It remains unclear whether glucocorticoid treatment can improve the outcome of sepsis. The aim of the present study was to investigate if glucocorticoid receptor (GR) expression and function is impaired in lipopolysaccharide (LPS) induced shock, and whether the time point for start of glucocorticoid treatment affects the outcome. Male C57BL/6J mice were administered LPS i.p. and GR expression and binding ability in blood and spleen leukocytes were analysed by flow cytometry. GR translocation was analysed using Image Stream technique. The effect of dexamethasone treatment started 2 h before or 2, 12 or 36 h after LPS administration on survival was studied. Despite increased GR expression in neutrophils after LPS administration, the GR binding capacity was reduced. In addition, GR translocation was decreased in neutrophils and T lymphocytes from endotoxic mice at 12 h compared to control animals. Dexamethasone treatment improved survival only when started early (2 h) after LPS administration. The decreased glucocorticoid responsiveness displayed by neutrophils, in combination with their increased numbers, may explain why survival is increased only when dexamethasone treatment is given early during LPS induced shock.
Product Description
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material