MilliporeSigma
  • Vizantin inhibits endotoxin-mediated immune responses via the TLR 4/MD-2 complex.
Vizantin inhibits endotoxin-mediated immune responses via the TLR 4/MD-2 complex.
Journal of immunology (Baltimore, Md. : 1950) (2014-09-28)
Masataka Oda, Hirofumi Yamamoto, Masahiro Shibutani, Mayo Nakano, Kenta Yabiku, Takafumi Tarui, Naoya Kameyama, Daiki Shirakawa, Sumiyo Obayashi, Naoyuki Watanabe, Hiroshi Nakase, Midori Suenaga, Yoichi Matsunaga, Masahiro Nagahama, Hironobu Takahashi, Hiroshi Imagawa, Mie Kurosawa, Yutaka Terao, Mugio Nishizawa, Jun Sakurai
ABSTRACT
Vizantin has immunostimulating properties and anticancer activity. In this study, we investigated the molecular mechanism of immune activation by vizantin. THP-1 cells treated with small interfering RNA for TLR-4 abolished vizantin-induced macrophage activation processes such as chemokine release. In addition, compared with wild-type mice, the release of MIP-1β induced by vizantin in vivo was significantly decreased in TLR-4 knockout mice, but not in TLR-2 knockout mice. Vizantin induced the release of IL-8 when HEK293T cells were transiently cotransfected with TLR-4 and MD-2, but not when they were transfected with TLR-4 or MD-2 alone or with TLR-2 or TLR-2/MD-2. A dipyrromethene boron difluoride-conjugated vizantin colocalized with TLR-4/MD-2, but not with TLR-4 or MD-2 alone. A pull-down assay with vizantin-coated magnetic beads showed that vizantin bound to TLR-4/MD-2 in extracts from HEK293T cells expressing both TLR-4 and MD-2. Furthermore, vizantin blocked the LPS-induced release of TNF-α and IL-1β and inhibited death in mice. We also performed in silico docking simulation analysis of vizantin and MD-2 based on the structure of MD-2 complexed with the LPS antagonist E5564; the results suggested that vizantin could bind to the active pocket of MD-2. Our observations show that vizantin specifically binds to the TLR-4/MD-2 complex and that the vizantin receptor is identical to the LPS receptor. We conclude that vizantin could be an effective adjuvant and a therapeutic agent in the treatment of infectious diseases and the endotoxin shock caused by LPS.