MilliporeSigma
  • Colocalization of cocaine- and amphetamine-regulated transcript with kisspeptin and neurokinin B in the human infundibular region.
Colocalization of cocaine- and amphetamine-regulated transcript with kisspeptin and neurokinin B in the human infundibular region.
PloS one (2014-08-02)
Katalin Skrapits, Beáta Á Borsay, László Herczeg, Philippe Ciofi, Stephen R Bloom, Mohammad A Ghatei, Waljit S Dhillo, Zsolt Liposits, Erik Hrabovszky
ABSTRACT
Kisspeptin (KP)- and neurokinin B (NKB)- synthesizing neurons of the hypothalamic arcuate nucleus play a pivotal role in the regulation of pulsatile gonadotropin-releasing hormone (GnRH) secretion. Unlike in rodents and sheep, the homologous KP and NKB neurons in the human infundibular region rarely express dynorphin- but often exhibit Substance P (SP) immunoreactivity, indicating remarkable species differences in the neurochemical phenotype of these neurons. In search for additional neuropeptides in human KP and NKB neurons, we carried out immunofluorescent studies on hypothalamic sections obtained from five postmenopausal women. Colocalization experiments provided evidence for the presence of cocaine- and amphetamine-regulated transcript (CART) in 47.9 ± 6.6% of KP-immunoreactive (IR) and 30.0 ± 4.9% of NKB-IR perikarya and in 17.0 ± 2.3% of KP-IR and 6.2 ± 2.0% of NKB-IR axon varicosities. All three neuropeptides were present in 33.3 ± 4.9% of KP-IR and 28.2 ± 4.6% of NKB-IR somata, respectively, whereas triple-labeling showed lower incidences in KP-IR (14.3 ± 1.8%) and NKB-IR (5.9 ± 2.0%) axon varicosities. CART-IR KP and NKB neurons established contacts with other peptidergic cells, including GnRH-IR neurons and also sent projections to the infundibular stalk. KP and NKB fibers with CART often contained SP as well, while being distinct from CART fibers co-containing the orexigenic peptide agouti-related protein. Presence of CART in human, but not rodent, KP and NKB neurons represents a new example of species differences in the neuropeptide repertoire of mediobasal hypothalamic KP and NKB neurons. Target cells, receptor sites and physiological significance of CART in the efferent communication of KP and NKB neurons in primates require clarification.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, BioUltra, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%