MilliporeSigma
  • Cathepsin K-mediated Notch1 activation contributes to neovascularization in response to hypoxia.
Cathepsin K-mediated Notch1 activation contributes to neovascularization in response to hypoxia.
Nature communications (2014-06-05)
Haiying Jiang, Xian Wu Cheng, Guo-Ping Shi, Lina Hu, Aiko Inoue, Yumiko Yamamura, Hongxian Wu, Kyosuke Takeshita, Xiang Li, Zhe Huang, Haizhen Song, Masashi Asai, Chang-Ning Hao, Kazumasa Unno, Teruhiro Koike, Yoshiharu Oshida, Kenji Okumura, Toyoaki Murohara, Masafumi Kuzuya
ABSTRACT
Cysteine proteases play important roles in pathobiology. Here we reveal that cathepsin K (CatK) has a role in ischaemia-induced neovascularization. Femoral artery ligation-induced ischaemia in mice increases CatK expression and activity, and CatK-deficient mice show impaired functional recovery following hindlimb ischaemia. CatK deficiency reduces the levels of cleaved Notch1 (c-Notch1), Hes1 Hey1, Hey2, vascular endothelial growth factor, Flt-1 and phospho-Akt proteins of the ischaemic muscles. In endothelial cells, silencing of CatK mimicked, whereas CatK overexpression enhanced, the levels of c-Notch1 and the expression of Notch downstream signalling molecules, suggesting CatK contributes to Notch1 processing and activates downstream signalling. Moreover, CatK knockdown leads to defective endothelial cell invasion, proliferation and tube formation, and CatK deficiency is associated with decreased circulating endothelial progenitor cells-like CD31(+)/c-Kit(+) cells in mice following hindlimb ischaemia. Transplantation of bone marrow-derived mononuclear cells from CatK(+/+) mice restores the impairment of neovascularization in CatK(-/-) mice. We conclude that CatK may be a potential therapeutic target for ischaemic disease.
Sigma-Aldrich
CA-074 Me, CA-074 Me, CAS 147859-80-1, is a cell-permeable analog of CA-074 that acts as an irreversible inhibitor of intracellular cathepsin B.
Sigma-Aldrich
E-64 Protease Inhibitor, The E-64 Protease Inhibitor, also referenced under CAS 66701-25-5, controls the biological activity of E-64 Protease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Sigma-Aldrich
Triciribine hydrate, ≥97% (HPLC)
Sigma-Aldrich
Cathepsin S Active human, recombinant, expressed in FreeStyle 293-F cells, ≥90% (SDS-PAGE)