MilliporeSigma
  • Biologic factors associated with tumor oxygenation are prognostic in patients with stage III esophageal cancer: long-term results.
Biologic factors associated with tumor oxygenation are prognostic in patients with stage III esophageal cancer: long-term results.
Anticancer research (2014-07-31)
Dirk Rades, Amira Bajrovic, Steven E Schild, Christoph Thorns, Ergin Kilic
ABSTRACT
Long-term results of a study investigating potential prognostic factors for treatment outcomes in patients with stage III esophageal cancer are presented. In 64 patients, the impact of tumor cell expression of erythropoietin (EPO) and erythropoietin-receptor (EPO-R) and ten additional factors (age, gender, performance status, tumor length, tumor stage (T-stage), nodes (N-stage), histology/grading, hemoglobin levels during radiotherapy, surgery) on survival and loco-regional control was evaluated up to 10 years following radio-chemotherapy. On multivariate analysis, improved survival was associated with low EPO-R expression (p=0.034) and hemoglobin levels during radiotherapy ≥ 12 g/dl (p=0.026). Low EPO expression was associated with survival on univariate (p=0.010) but not on multivariate analysis (p=0.42). On multivariate analysis, improved loco-regional control was significantly associated with hemoglobin levels during radiotherapy ≥ 12 g/dl (p<0.001). The long-term results confirm that hemoglobin levels during radiotherapy and tumor cell expression of EPO-R are significant prognostic factors in patients with locally advanced esophageal cancer.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, BioReagent, ≥36.0% in H2O (T)
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Citric acid trisodium salt, anhydrous, ≥98% (GC)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, 1 M in H2O
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Supelco
5-Fluorouracil, analytical standard
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
USP
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Cisplatin, European Pharmacopoeia (EP) Reference Standard