MilliporeSigma
  • A multivalent chimeric vaccine composed of Schistosoma mansoni SmTSP-2 and Sm29 was able to induce protection against infection in mice.
A multivalent chimeric vaccine composed of Schistosoma mansoni SmTSP-2 and Sm29 was able to induce protection against infection in mice.
Parasite immunology (2014-04-23)
C S Pinheiro, A P D Ribeiro, F C Cardoso, V P Martins, B C P Figueiredo, N R G Assis, S B Morais, M V Caliari, A Loukas, S C Oliveira
ABSTRACT
Schistosoma mansoni is a blood fluke parasite responsible for schistosomiasis. The best long-term strategy to control schistosomiasis is through immunization combined with drug treatment. In this study, we cloned, expressed and purified SmTSP-2 fused to the N- and C-terminal halves of Sm29 and tested these chimeras as vaccine candidates using an adjuvant approved to be used in humans. The results demonstrated that vaccination with SmTSP-2 fused to N- or C-terminus of Sm29-induced reduction in worm burden and liver pathology when compared to control animals. Additionally, we detected high levels of mouse-specific IgG, IgG1 and IgG2a against both chimeras and significant amounts of IFN-γ and TNF-α and no IL-4. Finally, studies with sera from patients resistant to infection and living in schistosomiasis endemic areas revealed high levels of specific IgG to both chimeras when compared to healthy individuals. In conclusion, SmTSP-2/Sm29 chimeras tested here induced partial protection against infection and might be a potential vaccine candidate.
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Bovine TNFα / Tumor Necrosis Factor alpha ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Mouse Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for cell and tissue lysates