MilliporeSigma
  • Tsc1-Tp53 loss induces mesothelioma in mice, and evidence for this mechanism in human mesothelioma.
Tsc1-Tp53 loss induces mesothelioma in mice, and evidence for this mechanism in human mesothelioma.
Oncogene (2013-07-16)
Y Guo, L R Chirieac, R Bueno, H Pass, W Wu, I A Malinowska, D J Kwiatkowski
ABSTRACT
Mesothelioma is diagnosed in ∼2500 patients in the United States every year, most often arising in the pleural space, but also occurring as primary peritoneal mesothelioma. The vast majority of patients with mesothelioma die of their disease within 3 years. We developed a new mouse model of mesothelioma by bladder or intraperitoneal injection of adenovirus Cre into mice with conditional alleles of each of Tp53 and Tsc1. Such mice began to develop malignant ascites about 6 months after injection, which was due to peritoneal mesothelioma, on the basis of tumor morphology and immunohistochemical staining. Mesothelioma cell lines were established, which showed loss of both Tsc1 and Tp53, with mammalian target of rapamycin complex (mTORC)1 activation. Treatment of mice with malignant ascites due to mesothelioma with rapamycin led to a marked reduction in ascites, extended survival and a 95-99% reduction in the mesothelioma tumor volume, in comparison with vehicle-treated mice. To see whether TSC1/TSC2 loss was a common genetic event in human mesothelioma, we examined nine human mesothelioma cell lines, and found that four of nine showed persistent activation of mTORC1, although none had loss of TSC1 or TSC2. A tissue microarray analysis of 198 human mesothelioma specimens showed that 33% of cases had reduced TSC2 expression and 60% showed activation of mTOR, indicating that mTOR activation is common in human mesothelioma, suggesting that it is a potential therapeutic target.
Product Description
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Anti-Cytokeratin AE1/AE3 Antibody, recognizes acidic & basic cytokeratins, clone AE1/AE3, clone AE1/AE3, Chemicon®, from mouse
Sigma-Aldrich
Hematoxylin, certified by the BSC
Millipore
Bifido Selective Supplement B, suitable for microbiology
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard