MilliporeSigma
  • Characterization of a plant-produced recombinant human secretory IgA with broad neutralizing activity against HIV.
Characterization of a plant-produced recombinant human secretory IgA with broad neutralizing activity against HIV.
mAbs (2014-12-09)
Matthew Paul, Rajko Reljic, Katja Klein, Pascal M W Drake, Craig van Dolleweerd, Martin Pabst, Markus Windwarder, Elsa Arcalis, Eva Stoger, Friedrich Altmann, Catherine Cosgrove, Angela Bartolf, Susan Baden, Julian K-C Ma
ABSTRACT
Recombinant Secretory IgA (SIgA) complexes have the potential to improve antibody-based passive immunotherapeutic approaches to combat many mucosal pathogens. In this report, we describe the expression, purification and characterization of a human SIgA format of the broadly neutralizing anti-HIV monoclonal antibody (mAb) 2G12, using both transgenic tobacco plants and transient expression in Nicotiana benthamiana as expression hosts (P2G12 SIgA). The resulting heterodecameric complexes accumulated in intracellular compartments in leaf tissue, including the vacuole. SIgA complexes could not be detected in the apoplast. Maximum yields of antibody were 15.2 μg/g leaf fresh mass (LFM) in transgenic tobacco and 25 μg/g LFM after transient expression, and assembly of SIgA complexes was superior in transgenic tobacco. Protein L purified antibody specifically bound HIV gp140 and neutralised tier 2 and tier 3 HIV isolates. Glycoanalysis revealed predominantly high mannose structures present on most N-glycosylation sites, with limited evidence for complex glycosylation or processing to paucimannosidic forms. O-glycan structures were not identified. Functionally, P2G12 SIgA, but not IgG, effectively aggregated HIV virions. Binding of P2G12 SIgA was observed to CD209 / DC-SIGN, but not to CD89 / FcalphaR on a monocyte cell line. Furthermore, P2G12 SIgA demonstrated enhanced stability in mucosal secretions in comparison to P2G12 IgG mAb.
Product Description
SAFC
BIS-TRIS
SAFC
BIS-TRIS
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Anti-Human IgA (α-chain specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Human IgA (α-chain specific)−Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
IgA from human colostrum, reagent grade, buffered aqueous solution