MilliporeSigma
  • Cord blood CD34+ cells expanded on Wharton's jelly multipotent mesenchymal stromal cells improve the hematopoietic engraftment in NOD/SCID mice.
Cord blood CD34+ cells expanded on Wharton's jelly multipotent mesenchymal stromal cells improve the hematopoietic engraftment in NOD/SCID mice.
European journal of haematology (2014-05-07)
Luisa Milazzo, Francesca Vulcano, Alessandra Barca, Giampiero Macioce, Emanuela Paldino, Stefania Rossi, Carmela Ciccarelli, Hamisa J Hassan, Adele Giampaolo
ABSTRACT
This study aims to investigate the capability of Wharton's jelly multipotent mesenchymal stromal cells (WJ-MSC) to support the in vitro expansion of hematopoietic stem/progenitor cells (HSPC) derived from cord blood (CB) in the absence of exogenous cytokines, and the effect on engraftment of the expanded cells in a mouse model. CB-CD34+ cells were seeded on WJ-MSC layer and cultured in HP01 serum-free medium. Day-7 and day-13 expanded cells were transplanted in NOD/SCID mice. After 8 wk, engraftment was evaluated in mouse bone marrow as percentage of human CD45+ cells. CD34+ population was expanded without increasing the differentiation rate. Co-culture increased the expansion of the CD34+ cells by 2.0 and 7.3 times after 7 and 13 d, respectively, and maintained the CD34+ cells up to day 20. In particular, earlier CD34+/CD90+ and CD34+/CD33- subtypes were increased. An advantage of the day-7 co-cultured HSPC in respect of HSPC at day 0 in the engraftment of NOD/SCID mice was obtained both as percentage of mice engrafted (100% vs. 75%) and as percentage of chimerism. Although the increase in hematopoietic progenitors is not dramatic as in the presence of added cytokines, this study demonstrates the effectiveness of the WJ-MSC not only to preserve the CD34+ population but also to improve the repopulating efficacy of the amplified HSPC, also in the absence of added cytokines and growth factors.
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source